Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM20B Antibody (1018512) [CoraFluor™ 1], Novus Biologicals™
SDP

Catalog No. FAB8427CL1 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8427CL1 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FAB8427CL1 Supplier Novus Biologicals Supplier No. FAB8427CL1
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FAM20B Monoclonal antibody specifically detects FAM20B in Human samples. It is validated for Western Blot

Specifications

Antigen FAM20B
Applications Western Blot
Classification Monoclonal
Clone 1018512
Conjugate CoraFluor 1
Formulation PBS
Gene Alias EC 2.7.1.-, family with sequence similarity 20, member B, glycosaminoglycan xylosylkinase, gxk1, KIAA0475, Protein FAM20B, Xylose kinase
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human FAM20B, Asn27-Leu409, Accession # O75063
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9917
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.