Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM20C Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982187
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2187 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2187 Supplier Bioss Supplier No. BS12376R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Calcium-binding protein which may play a role in dentin mineralization.

Specifications

Antigen FAM20C
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FAM20C
Gene Accession No. Q8IXL6
Gene Alias C76981; Dentin matrix protein 4; Dmp4; DMP-4; Extracellular serine/threonine protein kinase FAM20C; Fam20c; family with sequence similarity 20 member C; family with sequence similarity 20, member C; GEF-CK; Golgi casein kinase; Golgi-enriched fraction casein kinase; Kiaa4081; mKIAA4081; RGD1311980; RNS
Gene Symbols FAM20C
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FAM20C.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56975
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.