Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM212A Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305938F
Description
FAM212A Polyclonal specifically detects FAM212A in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
FAM212A | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
C3orf54, chromosome 3 open reading frame 54, family with sequence similarity 212, member A, INKA1, INKA1 protein, MGC20416 | |
Produced in rabbits immunized with E. coli-derived Human FAM212A fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
FITC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
389119 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction