Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM212A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559769
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559769 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559769 Supplier Invitrogen™ Supplier No. PA559769
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WDSGFSEVSG STWREEELPV SQRPAPSAQP LRRQCLSVSG LPMPSRAPVA SVPPVHHPRP KSTPDACLEH WQGLEAEDWT AALLNRGRS Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 76%.

FAM212A, initially identified as INCA (induced in neural crest by AP-2alpha) in xenopus, is expressed primarily in neural crest cells and their derivatives. FAM212A can associate with the P21-kinase protein PAK4 and this association modulates cytoskeletal dynamics. In mammals, FAM212A is thought to play a role in neural tube closure; a significant fraction of FAM212A-null mice exhibited exencephaly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM212A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM212A
Gene Accession No. Q96EL1
Gene Alias 6230427J02Rik; C3orf54; FAM212A; family with sequence similarity 212 member A; family with sequence similarity 212, member A; HInca; INCA; Induced in neural crest by AP2-alpha protein homolog; Inka box actin regulator 1; INKA1; INKA1 protein; MInca; PAK4-inhibitor INKA1; protein FAM212A; RGD1560778
Gene Symbols FAM212A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM212A. Recombinant protein control fragment (Product #RP-100269).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389119
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.