Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM212B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555698
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555698 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555698 Supplier Invitrogen™ Supplier No. PA555698
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EHPCWEGGRG PARPTVCSPS SQPSLGSSTK FPSHRSVCGR DLAPLPRTQP HQSCAQQGPE RVEPDDWTST LMSR Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 62%.

FAM212B (INKA2, Inka Box Actin Regulator 2) is a Protein Coding gene. An important paralog of this gene is ENSG00000284755. Inhibitor of the serine/threonine-protein kinase PAK4. Acts by binding PAK4 in a substrate-like manner, inhibiting the protein kinase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM212B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM212B
Gene Accession No. Q9NTI7
Gene Alias 6530418L21Rik; C1orf183; C3H1orf183; FAM212B; family with sequence similarity 212 member B; family with sequence similarity 212, member B; HInca-r; Induced in neural crest by AP2-alpha protein-related homolog; Inka box actin regulator 2; Inka2; MInca-r; PAK4-inhibitor INKA2; protein FAM212B; RGD1306526; uncharacterized protein C1orf183 homolog
Gene Symbols FAM212B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM212B. Recombinant protein control fragment (Product #RP-94549).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55924
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.