Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM212B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570353
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570353 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570353 Supplier Invitrogen™ Supplier No. PA570353
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 92%; Rat: 93%.

FAM212B (INKA2, Inka Box Actin Regulator 2) is a Protein Coding gene. An important paralog of this gene is ENSG00000284755. Inhibitor of the serine/threonine-protein kinase PAK4. Acts by binding PAK4 in a substrate-like manner, inhibiting the protein kinase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM212B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FAM212B
Gene Accession No. 0
Gene Alias 6530418L21Rik; C1orf183; C3H1orf183; FAM212B; family with sequence similarity 212 member B; family with sequence similarity 212, member B; HInca-r; Induced in neural crest by AP2-alpha protein-related homolog; Inka box actin regulator 2; Inka2; MInca-r; PAK4-inhibitor INKA2; protein FAM212B; RGD1306526; uncharacterized protein C1orf183 homolog
Gene Symbols FAM212B
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human FAM212B.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 310764
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.