Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM213A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552640
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552640 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552640 Supplier Invitrogen™ Supplier No. PA552640
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NTDVFLSKPQ KAALEYLEDI DLKTLEKEPR TFKAKELWEK NGAVIMAVRR PGCFLCREEA ADLSSLKSML DQLGVPLYAV VKEHIRTEVK DFQPYFKGE Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.

Involved in redox regulation of the cell (PubMed:26438880, PubMed:19951071). Acts as an antioxidant (PubMed:19951071, PubMed:26438880). Inhibits TNFSF11-induced NFKB1 and JUN activation and osteoclast differentiation (PubMed:19951071). May affect bone resorption and help to maintain bone mass (PubMed:19951071). Acts as a negative regulator of macrophage-mediated inflammation by inhibiting macrophage production of inflammatory cytokines, probably through suppression of the MAPK signaling pathway (PubMed:26438880). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM213A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM213A
Gene Accession No. Q6AXX6, Q9BRX8
Gene Alias C10orf58; Fam213a; family with sequence similarity 213 member A; family with sequence similarity 213, member A; Pamm; peroxiredoxin (PRX)-like 2 activated in M-CSF stimulated monocytes; peroxiredoxin-like 2 activated in M-CSF stimulated monocytes; Peroxiredoxin-like 2A; PRO2290; Protein PAMM; PRXL2A; PSEC0139; Redox-regulatory protein FAM213A; redox-regulatory protein PAMM; RGD1309676; Shp1; similar to RIKEN cDNA 5730469M10; sperm head protein 1; UNQ611/PRO1198; UPF0765 protein C10orf58; UPF0765 protein C10orf58 homolog
Gene Symbols FAM213A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM213A. Recombinant protein control fragment (Product #RP-90168).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361118, 84293
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.