Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM213B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569890
Description
This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 86%.
FAM213B is a protein coding gene.
Specifications
FAM213B | |
Polyclonal | |
Unconjugated | |
FAM213B | |
2810405K02Rik; AI836168; C16H1orf93; C1orf93; C5H1orf93; Fam213b; family with sequence similarity 213 member B; family with sequence similarity 213, member B; hypothetical protein MGC26818; LOW QUALITY PROTEIN: prostamide/prostaglandin F synthase; peroxiredoxin like 2B; Peroxiredoxin-like 2B; PM/PGFS; Prostamide/PG F synthase; prostamide/PGF synthase; prostamide/prostaglandin F synthase; Protein FAM213B; prxl2; Prxl2b; RGD1308251; sb:cb782; selu3; wu:fc33a02; zgc:85644 | |
Rabbit | |
Affinity chromatography | |
RUO | |
127281 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8TBF2 | |
FAM213B | |
synthetic peptide directed towards the middle region of human FAM213B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction