Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM213B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552290
Description
Immunogen sequence: LAGLLDQHGV RLVGVGPEAL GLQEFLDGDY FAGELYLDES KQLYKELGFK RYNSLSILPA ALGKPVRDVA AKAKAVGIQG NLSGDLLQSG GLLVVSKGGD KVLLHFVQKS Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 89%.
FAM213B is a protein coding gene.
Specifications
| FAM213B | |
| Polyclonal | |
| Unconjugated | |
| FAM213B | |
| 2810405K02Rik; AI836168; C16H1orf93; C1orf93; C5H1orf93; Fam213b; family with sequence similarity 213 member B; family with sequence similarity 213, member B; hypothetical protein MGC26818; LOW QUALITY PROTEIN: prostamide/prostaglandin F synthase; peroxiredoxin like 2B; Peroxiredoxin-like 2B; PM/PGFS; Prostamide/PG F synthase; prostamide/PGF synthase; prostamide/prostaglandin F synthase; Protein FAM213B; prxl2; Prxl2b; RGD1308251; sb:cb782; selu3; wu:fc33a02; zgc:85644 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 127281, 66469 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TBF2, Q9DB60 | |
| FAM213B | |
| Recombinant protein corresponding to Human FAM213B. Recombinant protein control fragment (Product #RP-88847). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction