Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM215A Monoclonal Antibody (6D11)
GREENER_CHOICE

Catalog No. PIMA521514
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521514 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521514 Supplier Invitrogen™ Supplier No. MA521514
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: KQKRKPASSE PTPEDALGGS AVPVRFHLHP EGLLWCSRCF FSHGPKGSEP PGRSAGLQGA TERSGRPSVQ AQAQACENLV PATVWDG.

The anti-Family with Sequence Similarity 215, Member A (Non-Protein Coding) (FAM215A) (AA 28-114) antibody from antibodies-online is a Mouse Monoclonal antibody to 6D11 to C17orf88, APR-2, FAM215A, family with sequence similarity 215, member A (non-protein coding), LINC00530.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM215A
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 6D11
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene FAM215A
Gene Accession No. Q9Y5M1
Gene Alias 2-Apr; APR-2; C17orf88; FAM215A; family with sequence similarity 215 member A (non-protein coding); LINC00530
Gene Symbols FAM215A
Host Species Mouse
Immunogen FAM215A (XP_498424, 28 a.a. ~ 114 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23591
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.