Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM217A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557277
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557277 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557277 Supplier Invitrogen™ Supplier No. PA557277
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SVDKQVGPYP GLPMPLGLCW PYADGDFFKN RNEIHVSSCS TIENNDGETL PAPNWNLKHG NSSVEENFTD ESDLSENEKT NDTLLSYFKK VDLNLKPETI K Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 70%.

FAM217A is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM217A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM217A
Gene Accession No. Q8IXS0
Gene Alias 1700026J04Rik; C35H6orf146; C6orf146; Fam217a; family with sequence similarity 217 member A; family with sequence similarity 217, member A; LOW QUALITY PROTEIN: protein FAM217A; protein FAM217A; uncharacterized protein C6orf146; uncharacterized protein C6orf146 homolog
Gene Symbols FAM217A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM217A. Recombinant protein control fragment (Product #RP-94876).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222826
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.