Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM217B Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298025R
Description
FAM217B Polyclonal specifically detects FAM217B in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
FAM217B | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
C20orf177, chromosome 20 open reading frame 177, dJ551D2.5, family with sequence similarity 217, member B, hypothetical protein LOC63939 | |
Produced in rabbits immunized with E. coli-derived Human FAM217B fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
63939 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction