Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM219A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565750
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565750 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565750 Supplier Invitrogen™ Supplier No. PA565750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GSPVNQQPKKN NVMARTRLVV PNKGYSSLDQ SPDEKPLVAL DTDSDDDFDM SRYSSSGYSS AE Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

The protein encoded by this gene has homologs that have been identified in mouse, macaque, etc organisms. Multiple alternatively spliced transcript variants that encode different protein isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM219A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM219A
Gene Accession No. Q8IW50
Gene Alias 2310028H24Rik; C11H9orf25; C9orf25; FAM219A; fam219aa; family with sequence similarity 219 member A; family with sequence similarity 219 member Aa; family with sequence similarity 219, member A; likely ortholog of H. sapiens chromosome 9 open reading frame 25 (C9orf25); protein FAM219A; uncharacterized protein C9orf25; uncharacterized protein C9orf25 homolog; zgc:101028
Gene Symbols FAM219A
Host Species Rabbit
Immunogen Recombinant Human FAM219A. Recombinant protein control fragment (Product #RP-106428).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 203259
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.