Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM219A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570109
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570109 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570109 Supplier Thermo Scientific Supplier No. PA570109
Add to Cart
Add to Cart

FAM219A Polyclonal Antibody

FAM219A Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM219A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FAM219A
Gene Accession No. Q9D772
Gene Alias 2310028H24Rik; C11H9orf25; C9orf25; FAM219A; fam219aa; family with sequence similarity 219 member A; family with sequence similarity 219 member Aa; family with sequence similarity 219, member A; likely ortholog of H. sapiens chromosome 9 open reading frame 25 (C9orf25); protein FAM219A; uncharacterized protein C9orf25; uncharacterized protein C9orf25 homolog; zgc:101028
Gene Symbols FAM219A
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human FAM219A (aa 49-98).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 71901
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.