Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM219B Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP326092MFV610
Description
FAM219B Polyclonal antibody specifically detects FAM219B in Human samples. It is validated for Western Blot, ImmunocytochemistrySpecifications
FAM219B | |
Polyclonal | |
Western Blot, Immunocytochemistry | |
C15orf17, chromosome 15 open reading frame 17, FLJ00005, hypothetical protein LOC57184 | |
Produced in rabbits immunized with E. coli-derived Human FAM219B fragment | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
57184 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction