Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM219B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571429
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571429 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571429 Supplier Invitrogen™ Supplier No. PA571429
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM219B Polyclonal Antibody for Western Blot

FAM219B (Family With Sequence Similarity 219 Member B) is a Protein Coding gene. An important paralog of this gene isFAM219A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM219B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FAM219B
Gene Accession No. Q5XKK7
Gene Alias C15orf17; FAM219B; family with sequence similarity 219 member B; family with sequence similarity 219, member B; protein FAM219B; uncharacterized protein C15orf17
Gene Symbols FAM219B
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human FAM219B.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57184
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.