Learn More
Invitrogen™ FAM219B Polyclonal Antibody

Description
Immunogen sequence: AKLQKHRDLA KAVLRRKGML GASPNRPDSS GKRSVKFNKG YTALSQSPDE NLVSLDSD Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.
Specifications
Specifications
| Antigen | FAM219B |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | FAM219B |
| Gene Accession No. | Q5XKK7 |
| Gene Alias | C15orf17; FAM219B; family with sequence similarity 219 member B; family with sequence similarity 219, member B; protein FAM219B; uncharacterized protein C15orf17 |
| Gene Symbols | FAM219B |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.