Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM21A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561604
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561604 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561604 Supplier Invitrogen™ Supplier No. PA561604
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VRVYDEEVEE PVLKAEAEKT EQEKTREQKE VDLIPKVQEA VNYGLQVLDS AFEQLDIKAG NSDSEEDDAN GRVELILEPK DLYIDRPLPY LIGSKLFME Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 91%.

Acts at least in part as component of the WASH core complex whose assembly at the surface of endosomes inhibits WASH nucleation-promoting factor (NPF) activity in recruiting and activating the Arp2/3 complex to induce actin polymerization and is involved in the fission of tubules that serve as transport intermediates during endosome sorting. Mediates the recruitment of the WASH core complex to endosome membranes via binding to phospholipids and VPS35 of the retromer CSC. Mediates the recruitment of the F-actin-capping protein dimer to the WASH core complex probably promoting localized F-actin polymerization needed for vesicle scission. Via its C-terminus binds various phospholipids, most strongly phosphatidylinositol 4-phosphate (PtdIns-(4)P), phosphatidylinositol 5-phosphate (PtdIns-(5)P) and phosphatidylinositol 3,5-bisphosphate (PtdIns-(3,5)P2). Involved in the endosome-to-plasma membrane trafficking and recycling of SNX27-retromer-dependent cargo proteins, such as GLUT1. Required for the association of DNAJC13, ENTR1, ANKRD50 with retromer CSC subunit VPS35. Required for the endosomal recruitment of CCC complex subunits COMMD1 and CCDC93 as well as the retriever complex subunit VPS35L. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM21A
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WASHC2A
Gene Accession No. Q641Q2
Gene Alias alternative protein FAM21B; bA56A21.1; bA98I6.1; FAM21A; FAM21B; family with sequence similarity 21 member A; family with sequence similarity 21, member A; family with sequence similarity 21, member B; WASH complex subunit 2A; WASH complex subunit FAM21A; WASH complex subunit FAM21B; WASHC2A
Gene Symbols WASHC2A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM21A. Recombinant protein control fragment (Product #RP-104434).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387680
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.