Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM21C Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988369
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8369 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8369 Supplier Bioss Supplier No. BS8214R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Component of the WASH complex, a complex present at the surface of endosomes that recruits and activates the Arp2/3 complex to induce actin polymerization. The WASH complex plays a key role in the fission of tubules that serve as transport intermediates during endosome sorting. In the complex, it probably mediates the recruitment of the complex to endosome membranes. Plays a role in fluid-phase endocytosis, a process exploited by vaccinia intracellular mature virus (IMV) to enter cells. As a result, may facilitate the penetration of IMV into cells.

Specifications

Antigen FAM21C
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene WASHC2C
Gene Accession No. Q9Y4E1
Gene Alias FAM21A; FAM21C; family with sequence similarity 21 member C; family with sequence similarity 21, member C; KIAA0592; Vaccinia virus penetration factor; VPEF; WASH complex subunit 2C; WASH complex subunit FAM21C; WASHC2C
Gene Symbols WASHC2C
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FAM21C.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253725
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.