Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM220A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565920
Description
Immunogen sequence: LEMRKDPSGAG LWLHSGGPV Highest antigen sequence identity to the following orthologs - mouse 50%, rat 55%.
May negatively regulate STAT3.
Specifications
FAM220A | |
Polyclonal | |
Unconjugated | |
FAM220A | |
ACPIN1; C7orf70; FAM220A; family with sequence similarity 220 member A; family with sequence similarity 220, member A; protein FAM220A; SIPAR; STAT3-interacting protein as a repressor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84792 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z4H9 | |
FAM220A | |
Recombinant Human FAM220A. Recombinant protein control fragment (Product #RP-106599). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction