Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM221A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555403
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555403 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555403 Supplier Invitrogen™ Supplier No. PA555403
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSPFLKAFQA SSSSSPETLT DVGTSSQVSS LRRPEEDDMA FFERRYQERM KMEKAAKWKG KAPLPSATK Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 67%.

Chromosome 7 has been linked to Osteogenesis imperfecta, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome. The deletion of a portion of the q arm of chromosome 7 is associated with Williams-Beuren syndrome, a condition characterized by mild mental retardation, an unusual comfort and friendliness with strangers and an elfin appearance. Deletions of portions of the q arm of chromosome 7 are also seen in a number of myeloid disorders including cases of acute myelogenous leukemia and myelodysplasia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM221A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM221A
Gene Accession No. A4D161
Gene Alias C7orf46; FAM221A; family with sequence similarity 221 member A; family with sequence similarity 221, member A; protein FAM221A; uncharacterized protein C7orf46
Gene Symbols FAM221A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM221A. Recombinant protein control fragment (Product #RP-92837).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340277
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.