Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM221B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571045
Description
This target displays homology in the following species: Human: 100%.
FAM221B is a protein coding gene.
Specifications
FAM221B | |
Polyclonal | |
Unconjugated | |
FAM221B | |
C9orf128; FAM221B; family with sequence similarity 221 member B; family with sequence similarity 221, member B; protein FAM221B; uncharacterized protein C9orf128 | |
Rabbit | |
Affinity chromatography | |
RUO | |
392307 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
A6H8Z2 | |
FAM221B | |
synthetic peptide directed towards the middle region of human FAM221B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction