Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM222A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5145112
Description
Immunogen sequence: QPLRAYSGST VASKSPEACG GRAYERASGS PLNCGVGLPT SFTVGQYFAA PWNSVLVTPT SDCYNPA Antigen sequence identity: Mouse 86%; Rat 84%.

Specifications
FAM222A | |
Polyclonal | |
Unconjugated | |
FAM222A | |
C12orf34; FAM222A; family with sequence similarity 222 member A; family with sequence similarity 222, member A; protein FAM222A | |
Rabbit | |
Affinity chromatography | |
RUO | |
84915 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5U5X8 | |
FAM222A | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 298-364. | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction