Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM222A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559093
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559093 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559093 Supplier Invitrogen™ Supplier No. PA559093
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSIHSLLYQL NQQCQAPGAA PPACQGMAIP HPSPAKHGPV PSFPSMAYSA AAGLPDCRKG TELGQGATQA LTLAGAAKPA GYADSGLDYL LWPQKP Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 85%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM222A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM222A
Gene Accession No. Q5U5X8
Gene Alias C12orf34; FAM222A; family with sequence similarity 222 member A; family with sequence similarity 222, member A; protein FAM222A
Gene Symbols FAM222A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM222A. Recombinant protein control fragment (Product #RP-97762).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84915
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.