Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM222B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113274
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113274 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113274 Supplier Invitrogen™ Supplier No. PA5113274
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM222B Polyclonal Antibody for Western Blot, IHC (P), ELISA

FAM222B is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM222B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene FAM222B
Gene Accession No. Q6P539, Q8WU58
Gene Alias 9630020I17Rik; C17orf63; FAM222B; family with sequence similarity 222 member B; family with sequence similarity 222, member B; protein FAM222B; uncharacterized protein C17orf63; uncharacterized protein C17orf63 homolog
Gene Symbols FAM222B
Host Species Rabbit
Immunogen Recombinant Human Protein FAM222B protein (351-562AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216971, 55731
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.