Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM227A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551817
Description
Immunogen sequence: TLVLKKPTQE VKRISEAREC ENMFPKKSCA ACKSPELTSN LFNIYGKSPL IVYFLQNYAS LQQHGKNVLI VRREKTTSTP DCTPTYTDVI SETLCSMKKR KDNLNQLYQH HWTEWNYFDK HLKELQDNFS REMKNIDPKA ADTK Highest antigen sequence identity to the following orthologs: Mouse - 49%, Rat - 49%.

Specifications
FAM227A | |
Polyclonal | |
Unconjugated | |
FAM227A | |
FAM227A; family with sequence similarity 227 member A; family with sequence similarity 227, member A; protein FAM227A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
646851 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
F5H4B4 | |
FAM227A | |
Recombinant protein corresponding to Human FAM227A. Recombinant protein control fragment (Product #RP-91523). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction