Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM228A Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145115
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145115 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145115 Supplier Invitrogen™ Supplier No. PA5145115
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FTFTSHCVIP KEWHKASARA RSKTYKYSPE KLIYADKKQK RKEKKTADLS QAAFERQFLS SKLSQKNKVG ERKGLVSRGL GRGWHAGLCS THE Antigen sequence identity: Mouse 35%; Rat 36%.

FAM228A (Family With Sequence Similarity 228 Member A) is a Protein Coding gene. Diseases associated with FAM228A include Echolalia. An important paralog of this gene is FAM228B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM228A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM228A
Gene Accession No. Q86W67
Gene Alias C2orf84; FAM228A; family with sequence similarity 228 member A; family with sequence similarity 228, member A; protein FAM228A; UPF0638 protein C2orf84
Gene Symbols FAM228A
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 107-199. Recombinant protein control fragment (Product #RP-110196).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 653140
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.