Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM234B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597896
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597896 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597896 Supplier Invitrogen™ Supplier No. PA597896
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM234B Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

FAM234B is a protein coding gene. Gene ontology (GO) annotation include integral component of membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM234B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene FAM234B
Gene Accession No. A2RU67
Gene Alias FAM234B; family with sequence similarity 234 member B; family with sequence similarity 234, member B; KIAA1467; Protein FAM234B
Gene Symbols FAM234B
Host Species Rabbit
Immunogen Recombinant Human Protein FAM234B protein (401-622AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57613
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.