Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM234B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582408
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582408 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582408 Supplier Invitrogen™ Supplier No. PA582408
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DLSPLELADV NGDGLRDVLL SFVMSRNGSA VGVSRPAANL VCLSGMNGST LWSSLLPEEA RDITCLELMP GSLAETICLV TGTHKMLSAF NATSGKAIWT LNPNYLSNGT LAAPVVVLPD LDEDGVR Highest antigen sequence indentity to the following orthologs: Rat - 83%, Mouse - 83%.

FAM234B is a protein coding gene. Gene ontology (GO) annotation include integral component of membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM234B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM234B
Gene Accession No. A2RU67
Gene Alias FAM234B; family with sequence similarity 234 member B; family with sequence similarity 234, member B; KIAA1467; Protein FAM234B
Gene Symbols FAM234B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM234B. Recombinant protein control fragment (Product #RP-89999).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57613
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.