Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM25A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP25979125UL
Description
FAM25A Polyclonal specifically detects FAM25A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FAM25A | |
Polyclonal | |
Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
BA96C23.5, Family With Sequence Similarity 25 Member A Family With Sequence Similarity 25, Member A, Protein FAM25A | |
Rabbit | |
Protein A purified | |
RUO | |
643161 | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2), containing 40% glycerol with 0.02% Sodium Azide | |
FAM25A | |
This antibody was developed against a recombinant protein corresponding to the amino acid sequence:EVVGHAKETGEKAIAEAIKKAQESGDKKMKEITETVTNTVTNAITHAAESL | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction