Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM26E Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565133
Description
Immunogen sequence: YAQKEKEQLEN TFLDYANKLS ERNLKCFFEN KRPDPFPMPT FAAWEAASEL HSFHQSQQHY STLHRVVDNG LQLSPEDDET TMVLVGTAHN M Highest antigen sequence identity to the following orthologs - mouse 74%, rat 77%.
Pore-forming subunit of a voltage-gated ion channel.
Specifications
FAM26E | |
Polyclonal | |
Unconjugated | |
CALHM5 | |
BB146404; C6orf188; Calcium homeostasis modulator protein 5; CALHM5; dJ493F7.3; Fam26e; family with sequence similarity 26 member E; family with sequence similarity 26, member E; protein FAM26E | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
254228 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N5C1 | |
CALHM5 | |
Recombinant Human FAM26E. Recombinant protein control fragment (Product #RP-105893). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction