Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM26F Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542606
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542606 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542606 Supplier Invitrogen™ Supplier No. PA542606
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: NRSCAAELPL VPCNQAKASD VQDLLKDLKA QSQVLGWILI AVVIIILLIF Sequence homology: Human: 100%; Rabbit: 92%; Rat: 92%.

Pore-forming subunit of a voltage-gated ion channel. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM26F
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CALHM6
Gene Accession No. Q5R3K3
Gene Alias C6orf187; Calcium homeostasis modulator protein 6; CALHM6; dJ93H18.5; Fam26f; family with sequence similarity 26 member F; family with sequence similarity 26, member F; LOC100353536; Protein FAM26F; RGD1304835
Gene Symbols CALHM6
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human FAM26F.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 441168
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.