Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM29A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989290
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9290 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9290 Supplier Bioss Supplier No. BS9723R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HAUS6 is 1 of 8 subunits of the 390-kDa human augmin complex, or HAUS complex. The augmin complex was first identified in Drosophila, and its name comes from the Latin verb augmentare, meaning to increase. The augmin complex is a microtubule-binding complex involved in microtubule generation within the mitotic spindle and is vital to mitotic spindle assembly (Goshima et al., 2008 ; Uehara et al., 2009 ).

Specifications

Antigen FAM29A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HAUS6
Gene Accession No. Q7Z4H7
Gene Alias 6230416J20Rik; 9830144E06; D4Ertd27e; DGT6; dim gamma-tubulin homolog; FAM29A; family with sequence similarity 29, member A; HAUS augmin like complex subunit 6; HAUS augmin-like complex subunit 6; HAUS augmin-like complex, subunit 6; HAUS6; KIAA1574; likely ortholog of H. sapiens family with sequence similarity 29, member A (FAM29A); mKIAA1574
Gene Symbols HAUS6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FAM29A/HAUS6.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54801
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.