Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM35A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP15550920UL
Description
FAM35A Polyclonal specifically detects FAM35A in Human samples. It is validated for Western Blot.Specifications
| FAM35A | |
| Polyclonal | |
| Western Blot 1:100-1:2000 | |
| Q86V20 | |
| FAM35A | |
| Synthetic peptides corresponding to FAM35A(family with sequence similarity 35, member A) Antibody(against the N terminal of FAM35A. Peptide sequence PDLSGHFLANCMNRHVHVKDDFVRSVSETQNIESQKIHSSRLSDITSSNM. | |
| Affinity Purified | |
| RUO | |
| 54537 | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| PBS and 2% Sucrose with 0.09% Sodium Azide | |
| bA163M19.1, FAM35A1, family with sequence similarity 35, member A, hypothetical protein LOC54537, MGC5560 | |
| Rabbit | |
| 94 kDa | |
| 20 μL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction