Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM35A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114103
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114103 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114103 Supplier Invitrogen™ Supplier No. PA5114103
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PDLSGHFLAN CMNRHVHVKD DFVRSVSETQ NIESQKIHSS RLSDITSSNM For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.

Component of the shieldin complex, which plays an important role in repair of DNA double-stranded breaks (DSBs) (PubMed:29656893, PubMed:29789392). During G1 and S phase of the cell cycle, the complex functions downstream of TP53BP1 to promote non-homologous end joining (NHEJ) and suppress DNA end resection (PubMed:29656893, PubMed:29789392). Mediates various NHEJ-dependent processes including immunoglobulin class-switch recombination, and fusion of unprotected telomeres (PubMed:29656893). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM35A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FAM35A
Gene Accession No. Q86V20
Gene Alias bA163M19.1; FAM35A; FAM35A1; family with sequence similarity 35 member A; family with sequence similarity 35, member A; protein FAM35A; RINN1-REV7-interacting novel NHEJ regulator 2; RINN2; Shield complex subunit 2; Shieldin complex subunit 2; SHLD2
Gene Symbols FAM35A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human FAM35A (aa 71-120).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54537
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.