Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM3A Monoclonal Antibody (358112)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA524107

Catalog No. PIMA524107


Only null left
Add to Cart

Description

Description

In direct ELISAs and Western blots, 100% cross-reactivity with recombinant mouse FAM3A is observed and no cross-reactivity with recombinant human (rh) FAM3B, rhFAM3C, or rhFAM3D is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

A novel cytokine-like gene family using structure-based methods to search for novel four-helix-bundle cytokines in genomics databases. There are four genes in this family, FAM3A, FAM3B, FAM3C, and FAM3D, each encoding a protein (224-235 amino acids) with a hydrophobic leader sequence. Northern analysis indicates that FAM3B is highly expressed in pancreas, FAM3D in placenta, and FAM3A and FAM3C in almost all tissues. Immunohistochemistry showed that FAM3A is expressed prominently in the vascular endothelium, particularly capillaries. Fam3a staining in capillary endothelium and endocardium of heart, as well as vascular endothelial staining in many other tissues.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FAM3A
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
P98173
FAM3A
Mouse myeloma cell line NS0-derived recombinant human FAM3A Phe34-Ser230.
100 μg
Primary
Human
Antibody
IgG2b
Western Blot
358112
Unconjugated
FAM3A
1810037C20Rik; 2.19; 2-19; Cytokine-like protein 2-19; DLD; DXS560S; FAM3 metabolism regulating signaling molecule A; FAM3A; family 3, member A protein; family with sequence similarity 3 member A; family with sequence similarity 3, member A; hypothetical protein MGC73350; protein FAM3A; RGD1562076; XAP-7; zgc:73350
Mouse
Protein A/G
RUO
60343
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.