Learn More
Invitrogen™ FAM3A Polyclonal Antibody

Description
The antibody detects endogenous levels of total FAM3A protein.
Specifications
Specifications
| Antigen | FAM3A |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | FAM3A |
| Gene Accession No. | P98173 |
| Gene Alias | 1810037C20Rik; 2.19; 2-19; Cytokine-like protein 2-19; DLD; DXS560S; FAM3 metabolism regulating signaling molecule A; FAM3A; family 3, member A protein; family with sequence similarity 3 member A; family with sequence similarity 3, member A; hypothetical protein MGC73350; protein FAM3A; RGD1562076; XAP-7; zgc:73350 |
| Gene Symbols | FAM3A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.