Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM3B Antibody [HRP], Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals AF2867H
Description
FAM3B Polyclonal antibody specifically detects FAM3B in Mouse samples. It is validated for Western BlotSpecifications
| FAM3B | |
| Polyclonal | |
| PBS | |
| Goat | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| HRP | |
| 2-21, C21orf11, C21orf76PANDER, chromosome 21 open reading frame 11, Cytokine-like protein 2-21, D21M16SJHU19e, family with sequence similarity 3, member B, ORF9, pancreatic derived factor, Pancreatic-derived factor, PRED44 | |
| Mouse myeloma cell line NS0-derived recombinant mouse FAM3B, Glu30-Arg235, Accession # Q9D309 | |
| 0.1 mL | |
| Cytokine Research | |
| 54097 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction