Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM3B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565050
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565050 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565050 Supplier Invitrogen™ Supplier No. PA565050
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AELIPDAPLSS AAYSIRSIGE RPVLKAPV Highest antigen sequence identity to the following orthologs - mouse 83%, rat 83%.

FAM3B (family with sequence similarity 3, member B), also known as ORF9, C21orf11 or PANDER, is a 235 amino acid secreted protein that localizes to discrete vesicular and perinuclear structures. Expressed at high levels in pancreas and at lower levels in kidney, colon, testis, prostate and small intestine, FAM3B functions as an islet-specific cytokine that promotes apoptosis and may inhibit Insulin secretion from beta-cells, possibly playing a role in overall pancreatic activity. Human FAM3B shares 78% sequence identity with its mouse counterpart, suggesting a conserved role between species. Three isoforms of FAM3B, designated A, B and C, exist due to alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM3B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM3B
Gene Accession No. P58499
Gene Alias 21-Feb; 2-21; 9030624C24Rik; C21orf11; C21orf76; cytokine-like protein 2-21; D16Jhu19e; D21M16SJHU19e; FAM3 metabolism regulating signaling molecule B; Fam3b; family with sequence similarity 3 member B; family with sequence similarity 3, member B; ORF9; pancreatic derived factor; pancreatic-derived factor; PANDER; PRED44; Protein FAM3B; RGD1311662; UNQ320/PRO365
Gene Symbols FAM3B
Host Species Rabbit
Immunogen Recombinant Human FAM3B. Recombinant protein control fragment (Product #RP-105814).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54097
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.