Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM3B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597732
Description
FAM3B Polyclonal Antibody for Western Blot, IHC (P), ELISA
FAM3B (family with sequence similarity 3, member B), also known as ORF9, C21orf11 or PANDER, is a 235 amino acid secreted protein that localizes to discrete vesicular and perinuclear structures. Expressed at high levels in pancreas and at lower levels in kidney, colon, testis, prostate and small intestine, FAM3B functions as an islet-specific cytokine that promotes apoptosis and may inhibit Insulin secretion from beta-cells, possibly playing a role in overall pancreatic activity. Human FAM3B shares 78% sequence identity with its mouse counterpart, suggesting a conserved role between species. Three isoforms of FAM3B, designated A, B and C, exist due to alternative splicing events.
Specifications
FAM3B | |
Polyclonal | |
Unconjugated | |
FAM3B | |
21-Feb; 2-21; 9030624C24Rik; C21orf11; C21orf76; cytokine-like protein 2-21; D16Jhu19e; D21M16SJHU19e; FAM3 metabolism regulating signaling molecule B; Fam3b; family with sequence similarity 3 member B; family with sequence similarity 3, member B; ORF9; pancreatic derived factor; pancreatic-derived factor; PANDER; PRED44; Protein FAM3B; RGD1311662; UNQ320/PRO365 | |
Rabbit | |
Protein G | |
RUO | |
304050, 52793, 54097 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
3 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
P58499, Q9D309 | |
FAM3B | |
Recombinant Human Protein FAM3B protein (31-235AA). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction