Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM3C Antibody (Delphy-1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197588
Description
FAM3C Monoclonal specifically detects FAM3C in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
FAM3C | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:1000, ELISA 1:100-1:2000, Immunoprecipitation 1:100 | |
FAM3C | |
Recombinant human ILEI (aa 42-227). | |
0.05 mg | |
Immunology | |
10447 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2a |
Western Blot, ELISA, Immunoprecipitation | |
Delphy-1 | |
Unconjugated | |
family with sequence similarity 3, member C, GS3876, ILEIInterleukin-like EMT inducer, predicted osteoblast protein | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction