Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM3D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109709
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109709 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109709 Supplier Invitrogen™ Supplier No. PA5109709
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RSYMSFSMKT IRLPRWLAAS PTKEIQVKKY KCGLIKPCPA NYFAFKICSG AANVVGPTMC FEDRMIMSPV KNNVGRGLNI ALVNGTTGAV LGQKAFDMYS GDVMHLVKFL KEIPGGALVL VASYDDPGTK MNDESRKLFS DLGSSYAKQL GFRDSWVFIG AKDLRGKSPF EQFLKNSPDT N.

FAM3D (FAM3 Metabolism Regulating Signaling Molecule D) is a Protein Coding gene. Diseases associated with FAM3D include Deafness, Autosomal Recessive 17 and Scheuermann Disease. Gene Ontology (GO) annotations related to this gene include cytokine activity. An important paralog of this gene is FAM3C. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM3D
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.11 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Fam3d
Gene Accession No. P97805, Q96BQ1
Gene Alias 2310076N21Rik; AV067083; cytokine-like protein EF-7; EF7; EF-7; Fam3d; family with sequence similarity 3 member D; family with sequence similarity 3, member D; Oit1; oncoprotein induced transcript 1; oncoprotein induced transcript 1 homolog; oncoprotein-induced protein 1; Protein EF-7; protein FAM3D; UNQ567/PRO1130
Gene Symbols Fam3d, Oit1
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 131177, 18300, 289949
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.