Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM3D Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | FAM3D |
---|---|
Concentration | 0.23 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-7905
![]() ![]() |
Proteintech Group Inc
123361AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Members of the FAM3 family were identified based on structural, but not sequence, homology to short chain cytokines including IL-2, IL-4 and GM-CSF. FAM3 proteins are four helix bundle cytokines with four conserved cysteines in all members (FAM3A-D). FAM3B (also known as PANDER, for pancreatic derived factor) is highly expressed in alpha and beta cells of the pancreas and is being investigated as a potential contributor to beta cell death and development of type I diabetes.Specifications
FAM3D | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P97805, Q96BQ1 | |
131177, 18300, 289949 | |
FAM3D Fusion Protein Ag2965 | |
Primary | |
-20°C | |
FAM3D |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
EF7, FAM3D, OIT1, Protein FAM3D | |
Fam3d | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title