Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM3D Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983349
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3349 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3349 Supplier Bioss Supplier No. BS14992R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Members of the FAM3 family were identified based on structural, but not sequence, homology to short chain cytokines including IL-2, IL-4 and GM-CSF. FAM3 proteins are four helix bundle cytokines with four conserved cysteines in all members (FAM3A-D). FAM3B (also known as PANDER, for pancreatic derived factor) is highly expressed in alpha and beta cells of the pancreas and is being investigated as a potential contributor to beta cell death and development of type I diabetes.

Specifications

Antigen FAM3D
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Fam3d
Gene Accession No. Q96BQ1
Gene Alias 2310076N21Rik; AV067083; cytokine-like protein EF-7; EF7; EF-7; FAM3D; family with sequence similarity 3 member D; family with sequence similarity 3, member D; Oit1; oncoprotein induced transcript 1; oncoprotein induced transcript 1 homolog; Oncoprotein-induced protein 1; Protein EF-7; Protein FAM3D; UNQ567/PRO1130
Gene Symbols Fam3d
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FAM3D.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 131177
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.