Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM40A Monoclonal Antibody (OTI7B8)
GREENER_CHOICE

Catalog No. PIMA525392
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525392 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525392 Supplier Invitrogen™ Supplier No. MA525392
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FAM40A Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

Plays a role in the regulation of cell morphology and cytoskeletal organization. Required in the cortical actin filament dynamics and cell shape.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM40A
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI7B8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene STRIP1
Gene Accession No. Q5VSL9
Gene Alias 6330569M22Rik; 6530401O14Rik; Fam40a; family with sequence similarity 40, member A; FAR11 factor arrest 11 homolog A; FAR11A; homolog of yeast FAR11 protein 1; hypothetical protein; Kiaa1761; Protein FAM40A; QnpA-18494; RP4-773N10.1; striatin interacting protein 1; striatin-interacting protein 1; striatin-interacting protein 1; LOW QUALITY PROTEIN: striatin-interacting protein 1; Strip1
Gene Symbols STRIP1
Host Species Mouse
Immunogen Full length human recombinant protein of FAM40A produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362012, 490117, 85369
Target Species Human, Rat, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.