Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM40A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA526722
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA526722 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA526722 Supplier Invitrogen™ Supplier No. PA526722
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine and non-human primate based on sequence homology.

Plays a role in the regulation of cell morphology and cytoskeletal organization. Required in the cortical actin filament dynamics and cell shape.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM40A
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene STRIP1
Gene Accession No. Q5VSL9, Q8C079
Gene Alias 6330569M22Rik; 6530401O14Rik; Fam40a; family with sequence similarity 40, member A; FAR11 factor arrest 11 homolog A; FAR11A; homolog of yeast FAR11 protein 1; hypothetical protein; Kiaa1761; Protein FAM40A; QnpA-18494; RP4-773N10.1; striatin interacting protein 1; striatin-interacting protein 1; striatin-interacting protein 1; LOW QUALITY PROTEIN: striatin-interacting protein 1; Strip1
Gene Symbols STRIP1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 352-381 amino acids from the central region of human FAM40A.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 229707, 85369
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.