Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM43A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113287
Description
FAM43A Polyclonal Antibody for Western Blot, IHC (P), ELISA
Specifications
| FAM43A | |
| Polyclonal | |
| Unconjugated | |
| FAM43A | |
| Fam43a; family with sequence similarity 43 member A; family with sequence similarity 43, member A; PP7298; Protein FAM43A; somitovasculin; Tuf1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 131583, 224093 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8BUP8, Q8N2R8 | |
| FAM43A | |
| Recombinant Human Protein FAM43A protein (1-200AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction