Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM44A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559504
Description
Immunogen sequence: KSEDTQKVKD EKQAKEKEVE SLKLPSEKNS NKAKTVEGTK EDFSLIDSDV DGLTDITVSS VHTSDLSSFE EDTEEEVVT Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 80%.

Specifications
FAM44A | |
Polyclonal | |
Unconjugated | |
BOD1L1 | |
biorientation of chromosomes in cell division 1 like 1; biorientation of chromosomes in cell division 1-like 1; biorientation of chromosomes in cell division protein 1-like 1; BOD1L; BOD1L1; FAM44A; family with sequence similarity 44, member A; KIAA1327 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
259282 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NFC6 | |
BOD1L1 | |
Recombinant protein corresponding to Human FAM44A. Recombinant protein control fragment (Product #RP-96593). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction