Learn More
Invitrogen™ FAM44B Polyclonal Antibody

Description
The antibody detects endogenous levels of total BOD1 protein.
Specifications
Specifications
| Antigen | FAM44B |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.6 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | BOD1 |
| Gene Accession No. | Q96IK1 |
| Gene Alias | 2310022M17Rik; biorientation defective 1; biorientation defective protein 1; biorientation of chromosomes in cell division 1; biorientation of chromosomes in cell division protein 1; BOD1; BOD2.1; bone length and organs 1; Fam44b; family with sequence similarity 44, member B; Protein FAM44B; RGD1305622 |
| Gene Symbols | BOD1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.