Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM45A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571395
Description
FAM45A Polyclonal Antibody for Western Blot
FAM45A is a protein coding gene.
Specifications
FAM45A | |
Polyclonal | |
Unconjugated | |
FAM45A | |
DENN domain-containing protein 10; DENND10; FAM45A; family with sequence similarity 45 member A; family with sequence similarity 45, member A; protein FAM45A | |
Rabbit | |
Affinity chromatography | |
RUO | |
404636 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8TCE6 | |
FAM45A | |
synthetic peptide directed towards the middle region of human FAM45A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction